Orders LinkContactUsLinkComponent
Powder funnels
120 mm
  HECH44002044
Powder funnels
  HECH44002044
Powder funnels
Powder funnels
120 mm
22,70 €
1 * 5 items
Powder funnels, made of PP, translucent. For decanting powdery materials. Also suitable for liquids. Cone angle 60°.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable
  • Funnel O-Ø:
    120 mm