Order Entry
Export
Orders LinkContactUsLinkComponent
 

Racks for freezer

15 (3×5), 50 mm, 424×140×274 mm
  RATI5400022
 :   RATIOLAB
Racks for freezer
  RATI5400022
 :  RATIOLAB
Racks for freezer

15 (3×5), 50 mm, 424×140×274 mm

 
142,40 €
   
1 * 1 items
 

For the storing of cryoboxes in freezer cabinets and chests.

  • Robust construction
  • Handles for convenient transport
  • Made of stainless steel with surfaces for perfect cleaning

 : Cryo racks are supplied with shelf sizes for any standard cryoboxes.

 
  • Compartments:
    15 (3×5)
  • W×D×H ext.:
    424×140×274 mm
  • Seq:
    04
  • Cryobox height:
    50 mm