Order Entry
Export
Orders LinkContactUsLinkComponent
 

Racks for freezer

16 (1×16), 32 mm, 140×140×589 mm
  RATI5400037
 :   RATIOLAB
Racks for freezer
  RATI5400037
 :  RATIOLAB
Racks for freezer

16 (1×16), 32 mm, 140×140×589 mm

 
144,80 €
   
1 * 1 items
 

For the storing of cryoboxes in freezer cabinets and chests.

  • Robust construction
  • Handles for convenient transport
  • Made of stainless steel with surfaces for perfect cleaning

 : Cryo racks are supplied with shelf sizes for any standard cryoboxes.

 
  • Compartments:
    16 (1×16)
  • W×D×H ext.:
    140×140×589 mm
  • Seq:
    15
  • Cryobox height:
    32 mm