Order Entry
Export
Orders LinkContactUsLinkComponent
Biohazard Bags

Biohazard Bags

Disposable waste bags, with indicator field, PP, 32 L, 500 pcs/pack, 800×600 mm
Catalog # RATI7002200
Supplier:   RATIOLAB
Biohazard Bags
Catalog # RATI7002200
Supplier:  RATIOLAB
Biohazard Bags
Biohazard Bags

Disposable waste bags, with indicator field, PP, 32 L, 500 pcs/pack, 800×600 mm

Price
358,52 €
Log in for availability
1 * 500 items
Product Details & Documents

These disposable waste bags are made from strong PP film with a broad seam at the bottom. Intended for the sterilisation and disposal of hazardous and contaminated labware.

  • Rectangle white indicator patch field turns into dark when autoclaved
  • Bags must not be completely closed while autoclaving

Available with biohazard sticker and safety imprint in English, German, French and Spanish language.

Specifications
  • Packed:
    500 pcs/pack
  • Capacity:
    32 L
  • Description:
    Disposable waste bags, with indicator field
  • L×W:
    800×600 mm
  • Material:
    PP
  • Seq:
    03