Biohazard Bags
Disposable waste bags, with indicator field, PP, 22 L, 500 pcs/pack, 780×400 mm
Catalog # RATI7002150
Supplier: RATIOLAB
Jump to:
Product Details & Documents
These disposable waste bags are made from strong PP film with a broad seam at the bottom. Intended for the sterilisation and disposal of hazardous and contaminated labware.
- Rectangle white indicator patch field turns into dark when autoclaved
- Bags must not be completely closed while autoclaving
Available with biohazard sticker and safety imprint in English, German, French and Spanish language.
Specifications
- Packed:500 pcs/pack
- Capacity:22 L
- Description:Disposable waste bags, with indicator field
- L×W:780×400 mm
- Material:PP
- Seq:02



