Order Entry
Export
Orders LinkContactUsLinkComponent
 

Nitromethane, Sigma-Aldrich®

Glass bottle, 100 mL
Catalog # SIAL73478-100ML
Flammable
Irritant
Supplier:   Merck
Nitromethane, Sigma-Aldrich®
Catalog # SIAL73478-100ML
Supplier:  Merck
CAS Number:  75-52-5
Nitromethane, Sigma-Aldrich®

Glass bottle, 100 mL

CAS Number:  75-52-5
Price
148,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Merck Index:13,06644
  • Boiling Pt:101.2 °C (1013 hPa)
  • Melting Pt:–29 °C
  • Density:1.138 g/cm³ (20 °C)
  • Flash Pt:44 °C
  • Storage Temperature:Ambient
  • UN:1261
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:200-876-6
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications