Methylcyclohexane, Sigma-Aldrich®
Glass bottle, 1 L
Catalog # SIAL300306-1L
Supplier: Merck
Jump to:
Product Details & Documents
- CAS Number:108-87-2
- Formula:C₆H₁₁CH₃
- MDL Number:MFCD00001497
- Molecular Weight:98.19 g/mol
- Merck Index:13,06074
- Boiling Pt:101 °C (1013 hPa)
- Melting Pt:–126 °C
- Density:0.7693 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- UN:2296
- ADR:3,II
- :M-Clarity™ quality level = MQ100
- EINECS:203-624-3
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



