Order Entry
Export
Orders LinkContactUsLinkComponent
n-Dodecane, Sigma-Aldrich®

n-Dodecane, Sigma-Aldrich®

Glass bottle, 1 L
Catalog # SIAL297879-1L
Health
Supplier:   Merck
n-Dodecane, Sigma-Aldrich®
Catalog # SIAL297879-1L
Supplier:  Merck
CAS Number:  112-40-3
n-Dodecane, Sigma-Aldrich®
n-Dodecane, Sigma-Aldrich®

Glass bottle, 1 L

CAS Number:  112-40-3
Price
883,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Dodecane
  • CAS Number:112-40-3
  • Formula:H₃C(CH₂)₁₀CH₃
  • MDL Number:MFCD00008969
  • Molecular Weight:170.34 g/mol
  • Boiling Pt:216 °C (1013 hPa)
  • Melting Pt:–9.6 °C
  • Density:0.7487 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Flash Pt:74 °C
  • Storage Temperature:Ambient
  • EINECS:203-967-9
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications