Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ninhydrin, Sigma-Aldrich®

Glass bottle for solids, 100 g
Catalog # SIAL151173-100G
Irritant
Supplier:   Merck
Ninhydrin, Sigma-Aldrich®
Catalog # SIAL151173-100G
Supplier:  Merck
CAS Number:  485-47-2
Ninhydrin, Sigma-Aldrich®
Ninhydrin, Sigma-Aldrich®

Glass bottle for solids, 100 g

CAS Number:  485-47-2
Price
188,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS Number:485-47-2
  • Formula:C₉H₆O₄
  • MDL Number:MFCD00003791
  • Molecular Weight:178.14 g/mol
  • Merck Index:13,06582
  • Boiling Pt:351 °C (1013 hPa)
  • Melting Pt:250…258 °C
  • Density:0.862 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:207-618-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications