Order Entry
Export
Orders LinkContactUsLinkComponent
Giemsa's stain modified solution, Sigma-Aldrich®

Giemsa's stain modified solution, Sigma-Aldrich®

Glass bottle, 250 mL
Catalog # 32884-250ML
Flammable
Toxic
Health
Giemsa's stain modified solution, Sigma-Aldrich®
Catalog # 32884-250ML
CAS Number:  51811-82-6
Giemsa's stain modified solution, Sigma-Aldrich®

Glass bottle, 250 mL

CAS Number:  51811-82-6
Price
38,90 €
Log in for availability
1 * 250 mL
Product Details & Documents

Modified Giemsa stain may be used for the identification of each human chromosome by differentials staining.

Modified Giemsa stain means when the pH of the Giemsa stain is changed from the usual 6.8 to 9.0.

Synonyms:  Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
  • CAS Number:51811-82-6
  • MDL Number:MFCD00081642
  • Density:0.99 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:257-438-2
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ200
  • Flash Pt:11 °C
  • UN:1230
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications