Giemsa's stain modified solution, Sigma-Aldrich®
Glass bottle, 1 L
Catalog # 32884-1L
Supplier: SIGMA ALDRICH MICROSCOPY
Giemsa's stain modified solution, Sigma-Aldrich®
Catalog # 32884-1L
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number: 51811-82-6
Jump to:
Product Details & Documents
Modified Giemsa stain may be used for the identification of each human chromosome by differentials staining.
Modified Giemsa stain means when the pH of the Giemsa stain is changed from the usual 6.8 to 9.0.
Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
- CAS Number:51811-82-6
- MDL Number:MFCD00081642
- Density:0.99 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:257-438-2
- ADR:3,II
- :M-Clarity™ quality level = MQ200
- Flash Pt:11 °C
- UN:1230
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



