Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
NMR STD/85% PHOSPHORIC ACID/D20(W/V) 5MM 1 * 1 items
NMR STD/85% PHOSPHORIC ACID/D20(W/V) 5MM 1 * 1 items
Print
NMR STD/85% PHOSPHORIC ACID/D20(W/V) 5MM 1 * 1 items
Catalog # SIAL612162-1EA
Supplier:
Merck
NMR STD/85% PHOSPHORIC ACID/D20(W/V) 5MM 1 * 1 items
Catalog # SIAL612162-1EA
Supplier:
Merck
CAS Number:
Expand image
Return to Top
NMR STD/85% PHOSPHORIC ACID/D20(W/V) 5MM 1 * 1 items
Add to cart
CAS Number:
Price
686,00 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Return Policy
Product Details & Documents