Capsaicin
Glass bottle for solids, 500 mg
Catalog # CAYM10010743-500
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 8-Methyl-N-vanillyl-6-none-amid
- CAS Number:404-86-4
- Formula:C₁₈H₂₇NO₃
- MDL Number:MFCD00017259
- Molecular Weight:305.42 g/mol
- Merck Index:12,01811
- Boiling Pt:210…220 °C (0.01 mmHg)
- Melting Pt:66.1 °C
- EINECS:206-969-8
- UN:2811
- ADR:6.1,III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



