Order Entry
Export
Orders LinkContactUsLinkComponent
 

Dye terminator removal, Mag-Bind® SeqDTR™

Mag-Bind™ SeqDTR™, 5 mL
Catalog # M1300-05
Supplier:   OMEGA BIO-TEK
Dye terminator removal, Mag-Bind® SeqDTR™
Catalog # M1300-05
Supplier:  OMEGA BIO-TEK
CAS Number:  
Dye terminator removal, Mag-Bind® SeqDTR™
Dye terminator removal, Mag-Bind® SeqDTR™

Mag-Bind™ SeqDTR™, 5 mL

CAS Number:  
Price
301,00 €
Log in for availability
1 * 5 mL
Product Details & Documents

Mag-Bind® SeqDTR™ is designed for the effective and reliable removal of unincorporated terminators from sequencing reaction. Sequencing products are first mixed with the Mag-Bind® SeqDTR™. DNA then selectively binds to the Mag-Bind® SeqDTR™ particles. With two rapid wash steps, trace contaminants such as nucleotides, primers and small, non targeted amplification products are removed.

Mag-Bind® SeqDTR™ can be processed in 96- and 384-well formats and is compatible with many liquid handling instruments.

Specifications

Specifications

  • Pk:
    5 mL
  • Description:
    Mag-Bind™ SeqDTR™
  • Cat. No.:
    M1300-05
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Target molecule:
    DNA
  • Method / Format:
    Magnetic beads
  • Description:
    Mag-Bind™ SeqDTR™
  • Sample type:
    PCR and other enzymatic reactions
  • Sample size:
    Variable

Specifications