Orders LinkContactUsLinkComponent
Microscale threaded basic kit, Kimble®
Microscale threaded basic kit, Kimble®
Micro vial, 3 ml
Catalog # SCERSP746100-0003
Supplier:   DWK Life Sciences
Microscale threaded basic kit, Kimble®
Catalog # SCERSP746100-0003
Supplier:  DWK Life Sciences
Microscale threaded basic kit, Kimble®
Micro vial, 3 ml
Price
81,10 €
Log in for availability
1 * 1 items
Product Details & Documents
This chemistry kit is designed for most common organic chemistry experiments.
Specifications
  • Description:
    Micro vial, 3 ml