Order Entry
Export
Orders LinkContactUsLinkComponent
Resistor PT100 1 * 1 items

Resistor PT100 1 * 1 items

Catalog # LAUDETP076
Supplier:   LAUDA
Resistor PT100 1 * 1 items
Catalog # LAUDETP076
Supplier:  LAUDA
Resistor PT100 1 * 1 items
Price
53,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Antikörper-Typ:
    Primär
  • Konjugation:
    Alexa Fluor® 350
  • Form:
    liquid
  • Reinigung:
    Purified by Protein A
  • Transporttemperatur:
    4°C
  • Reaktivität:
    Human|Rat|Mouse
  • Konzentration:
    1 μg/μl
  • Kreuzadsorption:
    No
  • ImmunoChemie:
    Yes
  • Cat. No.:
    BOSSBS-6456R-A350
  • Antigen-Name:
    Presenilin Enhancer Gamma Secretase Subunit
  • Lagertemperatur:
    Store at 4°C for 12 months
  • Gen-ID:
    55851
  • Immunogen:
    1-50/101
  • Antigen-Synonyme:
    PEN2|MDS033|Presenilin enhancer protein 2|Gamma-secretase subunit PEN-2|PEN-2|MSTP064
  • Lagerungspuffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Antigen-Symbol:
    PSENEN
  • Klonalität:
    Polyklonal
  • Isotyp:
    IgG
  • Wirt:
    Rabbit