Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Pipettes
Pipette Controllers
Pipetting pumps
Pipetting pumps
Print
Pipetting pumps
Pipetting pump for pipettes 5 - 10 ml, Green
Catalog # EDVO640
Supplier:
EDVOTEK
Pipetting pumps
Catalog # EDVO640
Supplier:
EDVOTEK
Expand image
Return to Top
Pipetting pumps
Pipetting pump for pipettes 5 - 10 ml, Green
Specifications
Add to cart
Jump to:
Specifications
Price
31,90 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Colour code:
Green
Description:
Pipetting pump for pipettes 5 - 10 ml