Order Entry
Export
Orders LinkContactUsLinkComponent
Eppendorf® Protein LoBind Plates

Eppendorf® Protein LoBind Plates

Microplate, White, 240 plates (10 bags x 24 plates), 140 µl
Catalog # 737-0238
Supplier:   EPPENDORF
Eppendorf® Protein LoBind Plates
Catalog # 737-0238
Supplier:  EPPENDORF
Eppendorf® Protein LoBind Plates
Eppendorf® Protein LoBind Plates

Microplate, White, 240 plates (10 bags x 24 plates), 140 µl

Price
2 628,00 €
Log in for availability
1 * 240 items
Product Details & Documents

Eppendorf Protein LoBind Plates are designed for use in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results.

  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimise the risk of sample interference
  • Lot-certified PCR clean purity grade: Free of human DNA, DNase, RNase and PCR inhibitors
  • High-contrast Unique OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors
  • RecoverMax® well design: Optimised well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing in plates

Protein sample recovery can be maximised with Eppendorf Protein LoBind Plates. Eppendorf LoBind plates are designed with a special, two-component polymer mix, creating a hydrophilic surface to ensure optimal sample recovery rates. These plates are specially developed for protein research or sensitive proteomic test methods, providing superior results by reducing sample binding to the plate surface.

Purity grade: PCR clean
Material: Polypropylene (PP)
Autoclavable: 121 °C, 20 min
Centrifugation stability: 6000×g
Well colour: Clear

Specifications
  • Packed:
    240 plates (10 bags x 24 plates)
  • Colour:
    White
  • Description:
    Microplate
  • Seq:
    09
  • Well volume:
    140 µl