Order Entry
Export
Orders LinkContactUsLinkComponent
Transport boxes, Securipack

Transport boxes, Securipack

5 L, Transport box for samples, load capacity 13,7 kg, 310×310×430 mm, 230×230×300 mm
Catalog # CORTFABRSECU5G1
Supplier:   E3 Cortex
Transport boxes, Securipack
Catalog # CORTFABRSECU5G1
Supplier:  E3 Cortex
Transport boxes, Securipack
Transport boxes, Securipack

5 L, Transport box for samples, load capacity 13,7 kg, 310×310×430 mm, 230×230×300 mm

Price
183,00 €
Log in for availability
1 * 1 items
Product Details & Documents

This Securipack packages are ready for use in the transport of samples of hazardous materials, except for Classes 7 and 6.2.

  • Very simple to use, this package eliminates risk posed by error or oversight
  • Mechanical high strength, in particular in the stacking by the juxtaposition of double walls on all the faces
  • High absorption power of the shocks on all the faces, guaranteeing a perfect protection in case of an accidental fall

Universal application for products regulated except for the radioactive and infectious products.

Protects against the following risks: It is a good oxidiser; non flammable and non toxic; corrosive

Ordering information: Supplied with 1 case (sheath), 2 crossed envelopes dressed in a foam thickness, 1 pocket of metallic or plastic retention and 1 sluggish absorbing product. Interior packaging and hazard class label may be supplied upon request.

Specifications
  • W×D×H ext.:
    310×310×430 mm
  • Capacity:
    5 L
  • Description:
    Transport box for samples, load capacity 13,7 kg
  • W×D×H int.:
    230×230×300 mm