Affinity chromatography columns, HiTrap™ MabSelect Xtra™
HiTrap™ MabSelect Xtra™, PP, 75 µm, 16×25 mm, 5×5 ml, <20 ml/min
Catalog # 28-4082-61
Supplier: Cytiva
Jump to:
Product Details & Documents
HiTrap™ MabSelect Xtra™ columns are prepacked with MabSelect Xtra™, a BioProcess medium for capture of monoclonal antbodies from large sample volumes.
- Increased dynamic binding capacity with smaller particle size and greater porosity than MabSelect™ medium
- MabSelect™ Xtra uses the same recombinant protein A ligand as MabSelect™
- Oriented coupling of recombinant protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
- Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
The increased capacity of the medium is well suited for purification of Mabs from high-level expression feedstock.
Documents
28408258_DATA FILE 1.pdf 28408258_DATA FILE.pdf 28408261_DATA FILE 1.pdf 28408261_DATA FILE.pdf
Datasheet
28408258_HAND BOOK 1.pdf 28408258_HAND BOOK.pdf 28408258_INSTRUCTIONS 1.pdf 28408258_INSTRUCTIONS 2.pdf 28408258_INSTRUCTIONS.pdf
Manual
More
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Flow rate:<20 ml/min
- Particle Size:75 µm
- Capacity:5×5 ml
- Description:HiTrap™ MabSelect Xtra™
- Seq:03
- Column hardware:PP
- Bed dimensions:16×25 mm
- Cat. No.:28-4082-61
- Max. pressure:5 bar
- pH range:3 - 10



