Order Entry
Export
Orders LinkContactUsLinkComponent
dNTP solutions, illustra™

dNTP solutions, illustra™

dATP, 100 mMol solution, 25 µmol
Catalog # 28-4065-01
Supplier:   Cytiva
dNTP solutions, illustra™
Catalog # 28-4065-01
Supplier:  Cytiva
CAS Number:  
dNTP solutions, illustra™
dNTP solutions, illustra™

dATP, 100 mMol solution, 25 µmol

CAS Number:  
Price
53,50 €
Log in for availability
1 * 25 µmol
Product Details & Documents

illustra™ dNTPs are high purity solutions for enzymatically sensitive applications, such as nucleic acid amplification, sequencing, labelling, mutagenesis, cDNA synthesis and expression profiling.

  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Specifications

Specifications

  • Pk:
    25 µmol
  • Description:
    dATP, 100 mMol solution

Specifications