ContactUsLinkComponent
Thermometer, Landsberger, mercury filled, measuring range: 17:35/0.05C, enclosed scale, capillary lens front, colourless, length:4 8 0mm, lower part: 180x6-7mm, 160mm immersion, Richter-top finish with button, suitable for government verification. 1 * 1 items
Catalog # AMARN63952
Supplier:  Amarell
Thermometer, Landsberger, mercury filled, measuring range: 17:35/0.05C, enclosed scale, capillary lens front, colourless, length:4 8 0mm, lower part: 180x6-7mm, 160mm immersion, Richter-top finish with button, suitable for government verification. 1 * 1 items
Catalog # AMARN63952
Supplier:  Amarell