Plugs for test tubes
LDPE, 16 mm, Yellow
Catalog # 212-0521
Supplier: BOETTGER PAUL
Jump to:
Product Details & Documents
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
- Not autoclavable
- Easy handling with a high, ribbed grip
- Easy to open by tilting 'one-hand operation'
Specifications
- Colour:Yellow
- Material:LDPE
- Seq:09
- Size:16 mm