Orders LinkContactUsLinkComponent
NMR tubes, polyethylene pressure caps
NMR tubes, polyethylene pressure caps
5 mm, Yellow
Catalog # SCERSP897095-0081
Supplier:   DWK Life Sciences
NMR tubes, polyethylene pressure caps
Catalog # SCERSP897095-0081
Supplier:  DWK Life Sciences
NMR tubes, polyethylene pressure caps
5 mm, Yellow
Price
34,30 €
Log in for availability
1 * 100 items
Product Details & Documents
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporisation pressures.
Specifications
  • Colour:
    Yellow
  • Length:
    5 mm
  • Seq:
    05