NMR tubes, polyethylene pressure caps
5 mm, Red
Catalog # SCERSP897095-0001
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporisation pressures.
Specifications
- Colour:Red
- Length:5 mm
- Seq:01