Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TISSUE GRINDER 1ML DUALL ALL GLASS 1ML 1 * 1 items
Catalog # SCERSP885452-0020
Supplier:
DWK Life Sciences
Expand image
TISSUE GRINDER 1ML DUALL ALL GLASS 1ML 1 * 1 items
Catalog # SCERSP885452-0020
Supplier:
DWK Life Sciences