Orders LinkContactUsLinkComponent
Homogeniser, Dounce type, KIMBLE®
7 ml
  432-0255
Homogeniser, Dounce type, KIMBLE®
  432-0255
Homogeniser, Dounce type, KIMBLE®
Homogeniser, Dounce type, KIMBLE®
7 ml
163,00 €
1 * 1 items
Designed primarily for cellular work where the nucleus remains intact after homogenisation.
  • All-glass construction
  • Two pestles are supplied with each complete unit
  • Large clearance pestle is used for the initial sample reduction
  • Small clearance pestle is used to form the final homogenate
  • Replacement components are available and completely interchangeable
  • Capacity:
    7 ml
  • Seq:
    15