Cork borer sets
Cork borer, 15,0 mm
217-0839
Brass, nickel plated, with handles and ejector.
- For manual boring of cork plugs
- Easy to clean
- Also available, cork drill sharpener compatible with all sets
- Hole Ø:15,0 mm
- Description:Cork borer
- Seq:12