Volumetric flasks, class A, blue markings, BLAUBRAND®
2000 ml, 29/32, 0,60 ± ml, Narrow mouth, with PP stopper
BRND2.37254
: Brand
Borosilicate glass 3.3, with PP stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- Total order from one batch, supplied with batch certificate
* In addition to DIN.
: DIN EN ISO 1042.
- NS:29/32
- Capacity:2000 ml
- Description:Narrow mouth, with PP stopper
- Seq:15
- Tolerance:0,60 ± ml