Disposable earplugs, Max®
Earplugs, without cord, refills for dispenser LS-400, Red, U*
Catalog # 111-0394
Supplier: HONEYWELL SAFETY
Jump to:
Product Details & Documents
Max® earplugs offer superior noise blocking performance in a disposable earplug.
- Bell shape provides maximum ear comfort
- Improved design is easier to insert
- Smooth, soil resistant covering prevents dirt penetrating the surface prior to insertion
SNR: 37 dB
SNR: 37 dB
EN 352-2:2002
Certifications: EN 352-2:2002
Specifications
- Colour:Red
- Description:Earplugs, without cord, refills for dispenser LS-400
- Seq:03
- Size:U*