Drum taps, brass
Self-closing with flame guard and PTFE seal, Viscous liquids >30 SAE, 2”
Catalog # MSGA2800
Supplier: asecos
Jump to:
Product Details & Documents
Brass taps for drums with screw thread.
- Self closing
- Flame guard
- PTFE seal
Ordering information: MSGA2799 includes a trailer fitting for decanting vessels.
Specifications
- Thread:2”
- Description:Self-closing with flame guard and PTFE seal
- Seq:03
- For:Viscous liquids >30 SAE