Drum taps, brass
Self-closing with flame guard, PTFE seal and trailer fitting, Non-corrosive liquids <30 SAE, 3/4"
Catalog # MSGA2799
Supplier: asecos
Jump to:
Product Details & Documents
Brass taps for drums with screw thread.
- Self closing
- Flame guard
- PTFE seal
Ordering information: MSGA2799 includes a trailer fitting for decanting vessels.
Specifications
- Thread:3/4"
- Description:Self-closing with flame guard, PTFE seal and trailer fitting
- Seq:01
- For:Non-corrosive liquids <30 SAE