Orders LinkContactUsLinkComponent
Drum taps, brass
Self-closing with flame guard and PTFE seal, Aggressive liquids, 3/4"
  MSGA2798
 :   asecos
Drum taps, brass
  MSGA2798
 :  asecos
Drum taps, brass
Drum taps, brass
Self-closing with flame guard and PTFE seal, Aggressive liquids, 3/4"
68,20 €
1 * 1 items
Brass taps for drums with screw thread.
  • Self closing
  • Flame guard
  • PTFE seal
 : MSGA2799 includes a trailer fitting for decanting vessels.
  • Thread:
    3/4"
  • Description:
    Self-closing with flame guard and PTFE seal
  • Seq:
    02
  • For:
    Aggressive liquids