7-Hydroxyindole 97%
Glass bottle, 1 g
Catalog # APOSOR10209-1G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:2380-84-9
- Formula:C₈H₇NO
- MDL Number:MFCD00152102
- Molecular Weight:133.15 g/mol
- Melting Pt:84…88 °C
- Storage Temperature:Freezer
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 g



