Order Entry
Export
Orders LinkContactUsLinkComponent
Imidazole ≥99%

Imidazole ≥99%

Glass bottle for solids, 500 g
Catalog # ABCRAB109053.0500
Health
Corrosive
Irritant
Supplier:   abcr
Imidazole ≥99%
Catalog # ABCRAB109053.0500
Supplier:  abcr
CAS Number:  288-32-4
Imidazole ≥99%
Imidazole ≥99%

Glass bottle for solids, 500 g

CAS Number:  288-32-4
Price
Inquire For Price
Log in for availability
1 * 500 g
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

Purity: 99% CAS No.: 288-32-4 Molecular Formula: C3H4N2 Formula Weight: 68.08 Flash Point: 145°(293°F) Physical Form: Cryst flakes Density: 1.03 Appearance: White to cream Melting Point: 88-92° Boiling Point: 256° Beilstein Reference: 103853 Merck Reference: 14,4912 UN No.: 2923 MDL No.: MFCD00005183

Excellent for buffer use in pH range 6.2 - 7.8. Also an histamine antagonist.

Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Merck Index:14,4912
  • Boiling Pt:256 °C
  • Melting Pt:88…92 °C
  • Density:1.03
  • Flash Pt:145 °C (293 °F)
  • Storage Temperature:Ambient
  • EINECS:206-019-2
  • UN:3263
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications