Order Entry
Export
Orders LinkContactUsLinkComponent
Calibrated Baumé hydrometers

Calibrated Baumé hydrometers

–2 - 15, 0,5, 315 mm
Catalog # ALLA0151TC002/20QP
Supplier:   Alla France
Calibrated Baumé hydrometers
Catalog # ALLA0151TC002/20QP
Supplier:  Alla France
Calibrated Baumé hydrometers
Calibrated Baumé hydrometers

–2 - 15, 0,5, 315 mm

Price
93,90 €
Log in for availability
1 * 1 items
Product Details & Documents

Baumé hydrometers calibrated at 20 °C.

Specifications
  • Length:
    315 mm
  • Seq:
    06
  • Division:
    0,5
  • Range:
    –2 - 15