Order Entry
Export
Orders LinkContactUsLinkComponent
 

Single channel pipettes, electronic, variable volume, Ovation®

Model for right handers, 5 - 250 µl, 1 µl, ±2,5 - ±0,8 %, <1,00 - <0,15 %, Blue
Catalog # VIST1065-0250
Single channel pipettes, electronic, variable volume, Ovation®
Catalog # VIST1065-0250
Single channel pipettes, electronic, variable volume, Ovation®
Single channel pipettes, electronic, variable volume, Ovation®

Model for right handers, 5 - 250 µl, 1 µl, ±2,5 - ±0,8 %, <1,00 - <0,15 %, Blue

Price
837,00 €
Log in for availability
1 * 1 items
Product Details & Documents

These pipettes are the ideal ergonomic choice for users who need a comprehensive range of automated liquid handling functions. An easy-to-read display and simple-to-operate keypad allows fast recall and setup of user-defined pipetting protocols and selection of operating modes, volumes and speeds.

  • Fully electronic operation maximises pipetting efficiency for tasks like multiple dispensing, pipetting, reverse pipetting, mixing, serial diluting and automatic sequential dispensing
  • Colour-coded right and left-handed models are available

Delivery information: These models require a power supply unit (sold separately) to recharge the pipette's battery.

Specifications
  • Colour code:
    Blue
  • Volume:
    5 - 250 µl
  • Description:
    Model for right handers
  • Seq:
    05
  • Increment:
    1 µl
  • Accuracy:
    ±2,5 - ±0,8 %
  • Imprecision:
    <1,00 - <0,15 %
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • VITON seal kit for Ovation 20 - 200 µl (QS), 101 - 200 µl (F1 and F2)
    VITON seal kit for Ovation 20 - 200 µl (QS), 101 - 200 µl (F1 and F2)
    VIST9057-2035
    Log in for availability
    1 * 1 KIT - 37,60 €

Product Information

  • Catalog #
    Availability
    Price
  • VITON seal kit for Ovation 100 - 1000 µl (QS), 201 - 1000 µl (F1 and F2)
    VITON seal kit for Ovation 100 - 1000 µl (QS), 201 - 1000 µl (F1 and F2)
    VIST9057-3036
    Log in for availability
    1 * 1 KIT - 37,60 €