Order Entry
Export
Orders LinkContactUsLinkComponent
Replacement Erlenmeyer flask caps, Corning®

Replacement Erlenmeyer flask caps, Corning®

Flat cap, 48 mm, for 2 L flask
Catalog # 734-4195
Supplier:   Corning
Replacement Erlenmeyer flask caps, Corning®
Catalog # 734-4195
Supplier:  Corning
Replacement Erlenmeyer flask caps, Corning®
Replacement Erlenmeyer flask caps, Corning®

Flat cap, 48 mm, for 2 L flask

Price
150,00 €
Log in for availability
1 * 24 items
Product Details & Documents

Made of PP.

Replacement caps for Corning® 500 ml, 2 and 3 litre Erlenmeyer flasks are available separately. They are supplied sterile and individually packaged.

Caution: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.

Specifications
  • Description:
    Flat cap, 48 mm, for 2 L flask
  • Seq:
    02