ContactUsLinkComponent
Vessel cell culture Roboflask for Automation maximum working volume 70ml cell growth area 92,6cm rectangular straight neck septum c ap TC-treated barcode sterile 1 * 100 items
Catalog # 734-4045
Supplier:  Corning
undefined
Vessel cell culture Roboflask for Automation maximum working volume 70ml cell growth area 92,6cm rectangular straight neck septum c ap TC-treated barcode sterile 1 * 100 items
Catalog # 734-4045
Supplier:  Corning