Order Entry
Export
Orders LinkContactUsLinkComponent
 

pH indicator, phenol red tablets and solutions, Lovibond®

Phenol red photometer tablet, —, MD100, MD200, MD600, MD610, Spectrodirect, MultiDirect, PM 600 and PM 620, 100 Tab.
Catalog # LOVI511770BT
Supplier:   Lovibond
pH indicator, phenol red tablets and solutions, Lovibond®
Catalog # LOVI511770BT
Supplier:  Lovibond
CAS Number:  
pH indicator, phenol red tablets and solutions, Lovibond®
pH indicator, phenol red tablets and solutions, Lovibond®

Phenol red photometer tablet, —, MD100, MD200, MD600, MD610, Spectrodirect, MultiDirect, PM 600 and PM 620, 100 Tab.

CAS Number:  
Price
25,90 €
Log in for availability
1 * 100 Tablet
Product Details & Documents

Indicator for pH range 6,5 to 8,4.

Can be used for measuring pH in conjunction with the Comparator 2000+ system.

 

Specifications

Specifications

  • Pk:
    100 Tab.
  • Description:
    Phenol red photometer tablet
  • For unit:
    MD100, MD200, MD600, MD610, Spectrodirect, MultiDirect, PM 600 and PM 620
  • Range:

Specifications