Order Entry
Export
Orders LinkContactUsLinkComponent
pH indicator, phenol red tablets and solutions, Lovibond®

pH indicator, phenol red tablets and solutions, Lovibond®

Phenol red tablet, 6,8 – 8,4 pH, Comparator 2000, 250 Tab.
Catalog # LOVI511751BT
Supplier:   Lovibond
pH indicator, phenol red tablets and solutions, Lovibond®
Catalog # LOVI511751BT
Supplier:  Lovibond
CAS Number:  
pH indicator, phenol red tablets and solutions, Lovibond®
pH indicator, phenol red tablets and solutions, Lovibond®

Phenol red tablet, 6,8 – 8,4 pH, Comparator 2000, 250 Tab.

CAS Number:  
Price
57,40 €
Log in for availability
1 * 250 Tablet
Product Details & Documents

Indicator for pH range 6,5 to 8,4.

Can be used for measuring pH in conjunction with the Comparator 2000+ system.

Documents

Specifications

Specifications

  • Pk:
    250 Tab.
  • Description:
    Phenol red tablet
  • For unit:
    Comparator 2000
  • Range:
    6,8 – 8,4 pH

Specifications