Orders LinkContactUsLinkComponent
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE keys for stopcocks, PYREX®, double bore, 2,5 mm
Catalog # BIBBSST203K
Supplier:   SciLabware
Spare PTFE Keys for Stopcocks, PYREX®
Catalog # BIBBSST203K
Supplier:  SciLabware
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE keys for stopcocks, PYREX®, double bore, 2,5 mm
Price
602,00 €
Log in for availability
1 * 10 items
Product Details & Documents
Fully interchangeable PTFE keys for Pyrex® stopcocks.
  • Supplied complete with plastic screw thread retaining device
  • Self lubricating, avoiding the need for stopcock grease
  • Made to a taper of 1:10 giving a combination of performance and ease of operation
 : Key is supplied complete with plastic screw thread retaining device.
Specifications
  • Hole Ø:
    2,5 mm
  • Description:
    Spare PTFE keys for stopcocks, PYREX®, double bore
  • Seq:
    04