Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STORAGE RACK FOR 20 SLIDES 1 * 1 items
Catalog # MEDT48-9051-00
Supplier:
MEDITE
Expand image
STORAGE RACK FOR 20 SLIDES 1 * 1 items
Catalog # MEDT48-9051-00
Supplier:
MEDITE