Orders LinkContactUsLinkComponent
Stoppers, universal caps, FitsAll™
Stoppers, universal caps, FitsAll™
Yellow, 12 - 16 mm
Catalog # SIMPT403Y
Supplier:   Simport Scientific
Stoppers, universal caps, FitsAll™
Catalog # SIMPT403Y
Supplier:  Simport Scientific
Stoppers, universal caps, FitsAll™
Stoppers, universal caps, FitsAll™
Yellow, 12 - 16 mm
Price
51,40 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Designed for easy-on, easy-off use. Cap removal is simple and quick. FitsAll™ closure is ideal for recapping most 12 and 16 mm O.D. evacuated glass blood collection tubes. Flexible FitsAll™ closures protect from aerosols of highly infectious microorganisms. They guard samples against cross-contamination and laboratory work areas against infection and spillage. Precision molded from low-density polyethylene, FitsAll™ fits firmly on the tube. Not suitable for autoclaving.
  • PE
  • Very sturdy under most rigorous procedures, including centrifugation
  • Available in 8 colours for easy sample identification
Specifications
  • Ø bottom:
    12 - 16 mm
  • Colour:
    Yellow