Inoculating loops and needles, Ino-Loop™
Inoculating loop, 1 µl, Blue, 25 loops/bag, 227 mm
Catalog # SIMPL200-1
Supplier: Simport Scientific
Jump to:
Product Details & Documents
PS, sterile. Loops and needles are smooth and flexible to facilitate uniform streaking without damaging the gel surface.
- Cross contamination due to improper sterilisation is eliminated
- Can be used under hoods without danger
- Colour-coded for ease of size identification
- Certificate of conformity available upon request
Needles are straight and suitable for removal of specimens of single colonies. Packed sterile in safe, tamperproof, zip-lock resealable bags.
Disposable inoculating loops and needles do not require flaming and thus eliminate the risk of infection due to aerosol formation of pathogenic substances.
Specifications
- Packed:25 loops/bag
- Capacity:1 µl
- Colour:Blue
- Description:Inoculating loop
- Length:227 mm
- Seq:02