ContactUsLinkComponent
Bottle pourer, PTFE, fits GL38 screw neck bottles, red ca
Catalog # COWI028.038RED
Supplier:  COWIE
Bottle pourer, PTFE, fits GL38 screw neck bottles, red ca
Catalog # COWI028.038RED
Supplier:  COWIE