Storage Bags
150×200 mm
Catalog # 300-0045
Supplier: Alpha Packaging
Jump to:
Product Details & Documents
Heavy duty storage bags, 500 gauge. Suitable for heat sealing. Clear PE, plain.
Specifications
- Dimensions:150×200 mm
- Seq:01
Heavy duty storage bags, 500 gauge. Suitable for heat sealing. Clear PE, plain.