Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
OBJECTIVE CFI Plan Achromat 10X Numerical Aperture: 0.25, Working Distance: 10.5 mm 1 * 1 items
Catalog # NIKOMRL00102
Supplier:
NIKON
Expand image
OBJECTIVE CFI Plan Achromat 10X Numerical Aperture: 0.25, Working Distance: 10.5 mm 1 * 1 items
Catalog # NIKOMRL00102
Supplier:
NIKON