Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium hydroxide ≥97.0%, shot, TECHNICAL

Sodium hydroxide ≥97.0%, shot, TECHNICAL

Plastic bucket, 25 kg
Catalog # 28240.460
Corrosive
Supplier:   VWR Chemicals
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Catalog # 28240.460
Supplier:  VWR Chemicals
CAS Number:  1310-73-2
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Sodium hydroxide ≥97.0%, shot, TECHNICAL

Plastic bucket, 25 kg

CAS Number:  1310-73-2
Price
282,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Specifications

Specification Test Results

  • Assay:
    Min. 97.0 %
  • Sodium carbonate:
    Max. 1.5 %

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg

Specifications