Orders LinkContactUsLinkComponent
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE keys for stopcocks, PYREX®, T-bore, 4,0 mm, Pyrex® stopcocks
Catalog # BIBBSST304K
Supplier:   SciLabware
Spare PTFE Keys for Stopcocks, PYREX®
Catalog # BIBBSST304K
Supplier:  SciLabware
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE Keys for Stopcocks, PYREX®
Spare PTFE keys for stopcocks, PYREX®, T-bore, 4,0 mm, Pyrex® stopcocks
Price
75,70 €
Log in for availability
1 * 5 items
Product Details & Documents
Fully interchangeable PTFE keys for Pyrex® stopcocks.
  • Supplied complete with plastic screw thread retaining device
  • Self lubricating, avoiding the need for stopcock grease
  • Made to a taper of 1:10 giving a combination of performance and ease of operation
Delivery information: Key is supplied complete with plastic screw thread retaining device.
Specifications
  • For:
    Pyrex® stopcocks
  • Hole Ø:
    4,0 mm
  • Description:
    Spare PTFE keys for stopcocks, PYREX®, T-bore
  • Seq:
    06